Assay Details
Gene Symbol: Gm17689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167584.1 | NP_001161056.1 | 2 - 3 | 204 | 72 | |
GenBank mRNA | EU703627.1 | - | 2 - 3 | 204 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
