Assay Details
Gene Symbol: Olfr1163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146644.2 | NP_666855.1 | 1 | 850 | 101 | |
GenBank mRNA | BC120531.1 | - | 1 | 850 | 40129454 | 101 |
BC120557.1 | - | 1 | 850 | 40129480 | 101 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1954788 | NM_146644.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1954788 | BC120531.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1954788 | BC120557.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126709 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1954788 | NM_146644.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1954788 | BC120531.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1954788 | BC120557.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P243393 | NM_146644.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P243393 | BC120531.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P243393 | BC120557.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P243393 | NM_146644.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P243393 | BC120531.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P243393 | BC120557.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408378 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408438 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408384 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408416 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21408446 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408415 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408378 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408409 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408412 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408413 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408404 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408395 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408426 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408425 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408392 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408443 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408385 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408397 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408441 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408406 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408431 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408428 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408388 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408384 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01735377 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408386 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01459875 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408382 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408400 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408430 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01529604 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408408 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408393 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408436 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408424 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408419 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408390 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408434 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408379 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408438 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408416 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408432 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01606968 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21408410 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10484715 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10484713 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10484712 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10484714 | - | - |
Affymetrix Mouse Clariom S Array | PSR0200033918.mm.1 | NM_146644.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200033918.mm.1 | NM_146644.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200033918.mm.1 | TC0200004006.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2970128 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002970128 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220328 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1220328 | - | - |
mouse-6 v1.1 (Illumina) | scl19037.1.1_224-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2970128 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl19037.1.1_224-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2970128 | - | - |
