Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200034078.mm.1 |
NM_146966.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034078.mm.1 |
NM_146966.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034078.mm.1 |
TC0200004065.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200034080.mm.1 |
NM_146966.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034080.mm.1 |
NM_146966.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034080.mm.1 |
TC0200004065.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200034079.mm.1 |
NM_146966.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034079.mm.1 |
NM_146966.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034079.mm.1 |
TC0200004065.mm.1 |
MTA TC |