Assay Details
| Gene Symbol: Olfr1270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146985.2 | NP_667196.1 | 1 | 652 | 95 | |
| GenBank mRNA | BC119113.1 | - | 1 | 652 | 8733862 | 95 |
| BC119115.1 | - | 1 | 652 | 8733864 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr1270
Genomic Map