Assay Details
| Gene Symbol: Amica1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005421.4 | NP_001005421.3 | 2 - 3 | 206 | 86 | |
| GenBank mRNA | CJ139627.1 | - | 2 - 3 | 160 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001005421.4 transcripts.
Set Membership: |
|

Amica1
Genomic Map