Assay Details
| Gene Symbol: Amica1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005421.4 | NP_001005421.3 | 7 - 8 | 1005 | 100 | |
| GenBank mRNA | AA120429.1 | - | 1 - 2 | 141 | 100 | |
| AY093688.1 | - | 7 - 8 | 1005 | 100 | ||
| BC050133.1 | - | 5 - 6 | 892 | 30023491 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Amica1
Genomic Map