Assay Details
| Gene Symbol: Amica1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005421.4 | NP_001005421.3 | 9 - 10 | 1224 | 110 | |
| GenBank mRNA | AA120429.1 | - | 3 - 4 | 360 | 110 | |
| AY093688.1 | - | 9 - 10 | 1224 | 110 | ||
| BC050133.1 | - | 7 - 8 | 1111 | 30023491 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Amica1
Genomic Map