Assay Details
| Gene Symbol: Olfr231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005520.2 | NP_001005520.2 | 1 | 651 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2134381 | NM_001005520.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2134381 | NM_001005520.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P128913 | NM_001005520.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20840927 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00254462 | - | - |
| Whole Mouse Genome G4122A | A_51_P128913 | NM_001005520.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00254462 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01099332 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00067565 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00067564 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01099327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20840929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00048382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00048380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00063661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01099324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00608570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01144036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00067564 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00067547 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00067538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01099320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00067542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01099329 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01099336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00681715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01099337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00067566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01099327 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20840926 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10360411 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10360410 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100029614.mm.1 | NM_001005520.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029614.mm.1 | NM_001005520.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029614.mm.1 | TC0100003600.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3163040 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3163040 | - | - |
| mouse-6 v1.1 (Illumina) | GI_53850665-A | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3163040 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3163040 | - | - |

Olfr231
Genomic Map