Assay Details
| Gene Symbol: Teddm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008426.3 | NP_001008426.1 | 1 | 255 | 82 | |
| GenBank mRNA | AK033834.1 | - | 1 | 256 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Teddm1b
Genomic Map