| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000023464.mm.1 |
NM_001150749.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023464.mm.1 |
NM_001150749.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023464.mm.1 |
NM_017473.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023464.mm.1 |
NM_017473.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023464.mm.1 |
TC1000003123.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000023463.mm.1 |
NM_001150749.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023463.mm.1 |
NM_001150749.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023463.mm.1 |
NM_017473.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023463.mm.1 |
NM_017473.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023463.mm.1 |
TC1000003123.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000023462.mm.1 |
NM_001150749.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023462.mm.1 |
NM_001150749.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023462.mm.1 |
NM_017473.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023462.mm.1 |
NM_017473.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023462.mm.1 |
TC1000003123.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000023465.mm.1 |
NM_001150749.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023465.mm.1 |
NM_001150749.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023465.mm.1 |
NM_017473.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023465.mm.1 |
NM_017473.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023465.mm.1 |
TC1000003123.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000023460.mm.1 |
NM_001150749.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023460.mm.1 |
NM_001150749.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023460.mm.1 |
NM_017473.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023460.mm.1 |
NM_017473.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023460.mm.1 |
TC1000003123.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000023461.mm.1 |
NM_001150749.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023461.mm.1 |
NM_001150749.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023461.mm.1 |
NM_017473.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023461.mm.1 |
NM_017473.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023461.mm.1 |
TC1000003123.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000023466.mm.1 |
NM_001150749.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023466.mm.1 |
NM_001150749.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023466.mm.1 |
NM_017473.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023466.mm.1 |
NM_017473.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023466.mm.1 |
TC1000003123.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000023471.mm.1 |
NM_001150749.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023471.mm.1 |
NM_001150749.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000023471.mm.1 |
TC1000003123.mm.1 |
MTA TC |