Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200009330.mm.1 |
NM_001004066.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009330.mm.1 |
NM_001286214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009330.mm.1 |
NM_019565.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009330.mm.1 |
TC1200001296.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009334.mm.1 |
NM_001004066.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009334.mm.1 |
TC1200001296.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009329.mm.1 |
NM_001004066.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009329.mm.1 |
NM_001286214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009329.mm.1 |
NM_019565.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009329.mm.1 |
TC1200001296.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009332.mm.1 |
NM_001004066.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009332.mm.1 |
NM_001286214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009332.mm.1 |
NM_019565.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009332.mm.1 |
TC1200001296.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009326.mm.1 |
NM_001004066.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009326.mm.1 |
NM_001286214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009326.mm.1 |
NM_019565.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009326.mm.1 |
TC1200001296.mm.1 |
MTA TC |