Assay Details
| Gene Symbol: Nxt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019761.6 | NP_062735.4 | 1 - 2 | 224 | 86 | |
| GenBank mRNA | AF156958.2 | - | 1 - 2 | 165 | 86 | |
| AK003254.1 | - | 1 - 2 | 200 | 86 | ||
| AK168314.1 | - | 1 - 2 | 184 | 86 | ||
| BC023672.1 | - | 1 - 2 | 182 | 5039139 | 86 | |
| CJ186513.1 | - | 1 - 2 | 213 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF156958.2, AK003254.1, AK168314.1, BC023672.1, NM_019761.6 transcripts.
Set Membership: |
|

Nxt1
Genomic Map