Assay Details
| Gene Symbol: Chst5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019950.2 | NP_064334.1 | 1 - 2 | 70 | 90 | |
| GenBank mRNA | AF176840.1 | - | 1 - 2 | 93 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF176840.1, NM_019950.2 transcripts.
Set Membership: |
|

Chst5
Genomic Map