Assay Details
| Gene Symbol: Olfr198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011808.1 | NP_001011808.1 | 1 | 549 | 197 | |
| GenBank mRNA | BC147215.1 | - | 1 | 644 | 9056251 | 197 |
More Information
Updated Mapping Information
Set Membership: |
|

Olfr198
Genomic Map