Assay Details
Gene Symbol: Olfr748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011837.2 | NP_001011837.2 | 1 | 645 | 102 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353569 | NM_001011837.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353569 | NM_001011837.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353569 | NM_001011837.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353569 | NM_001011837.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353569 | NM_001011837.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353569 | NM_001011837.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598115 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598105 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598113 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598122 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598141 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598106 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598127 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598124 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598123 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598120 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598137 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598116 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598138 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598126 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598109 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598118 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29598139 | - | - |
Whole Mouse Genome G4122A | A_51_P353569 | NM_001011837.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P353569 | NM_001011837.2 | mouse_transcriptome |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11140162 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11558582 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11140161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29598118 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598145 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11577030 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598105 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598108 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598112 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598116 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598111 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598126 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598142 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598103 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598123 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598140 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598150 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598118 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598139 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598141 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598147 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598137 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598106 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11734594 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598155 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598113 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598136 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598109 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598127 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598124 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598120 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29598122 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10414494 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10414493 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3160239 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_3160239 | - | - |
mouse-6 v1.1 (Illumina) | GI_58801435-S | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3160239 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3160239 | - | - |
