Assay Details
| Gene Symbol: G6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033221.3 | NP_001028393.1 | 1 - 2 | 76 | 100 | |
| NM_001191012.1 | NP_001177941.1 | 1 - 2 | 76 | 100 | ||
| GenBank mRNA | AK137516.1 | - | 1 - 2 | 77 | 100 | |
| GU269270.1 | - | 1 - 2 | 62 | 100 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

G6b
Genomic Map