Assay Details
| Gene Symbol: Zfp583 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033249.3 | NP_001028421.1 | 1 - 2 | 368 | 108 | |
| GenBank mRNA | AK133380.1 | - | 1 - 2 | 370 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK133380.1, NM_001033249.3 transcripts.
Set Membership: |
|

Zfp583
Genomic Map