Assay Details
| Gene Symbol: Mageb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033492.2 | NP_001028664.2 | 89 | 102 | ||
| GenBank mRNA | AK144342.1 | - | 90 | 102 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK144342.1,NM_001033492.2 transcripts.
This assay is in the 5'UTR region of the AK144342.1,NM_001033492.2 transcripts.
Set Membership: |
|

Mageb4
Genomic Map