Assay Details
| Gene Symbol: Mageb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033492.2 | NP_001028664.2 | 177 | 68 | ||
| GenBank mRNA | AK144342.1 | - | 178 | 68 | ||
| AK144350.1 | - | 143 | 68 | |||
| AK165797.1 | - | 161 | 68 | |||
| BC150912.1 | - | 122 | 9087823 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK144342.1, AK144350.1, BC150912.1, NM_001033492.2 transcripts.
Set Membership: |
|

Mageb4
Genomic Map