Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700001508.mm.1 |
TC0700000214.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700001508.mm.1 |
NM_001033638.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001508.mm.1 |
NM_001033638.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001508.mm.1 |
NM_001145190.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001508.mm.1 |
NM_001145190.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001499.mm.1 |
TC0700000214.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700001499.mm.1 |
NM_001033638.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001499.mm.1 |
NM_001033638.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001499.mm.1 |
NM_001205274.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001499.mm.1 |
NM_001205274.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001497.mm.1 |
TC0700000214.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700001497.mm.1 |
NM_001033638.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001497.mm.1 |
NM_001033638.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001497.mm.1 |
NM_001205274.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001497.mm.1 |
NM_001205274.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001498.mm.1 |
TC0700000214.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700001498.mm.1 |
NM_001033638.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001498.mm.1 |
NM_001033638.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001498.mm.1 |
NM_001205274.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700001498.mm.1 |
NM_001205274.1 |
Refseq |