Assay Details
| Gene Symbol: Pkig | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039391.2 | NP_001034480.1 | 1 - 2 | 107 | 158 | |
| NM_001277096.1 | NP_001264025.1 | 1 - 2 | 107 | 158 | ||
| GenBank mRNA | AK135505.1 | - | 1 - 2 | 108 | 158 | |
| CJ051558.1 | - | 1 - 2 | 108 | 158 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001039391.2,NM_001277096.1 transcripts.
This assay is in the 5'UTR region of the NM_001039391.2,NM_001277096.1 transcripts.
Set Membership: |
|

Pkig
Genomic Map