Assay Details
| Gene Symbol: Nfyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048168.2 | NP_001041633.1 | 6 - 7 | 714 | 95 | |
| NM_001277095.1 | NP_001264024.1 | 5 - 6 | 673 | 95 | ||
| NM_008692.4 | NP_032718.2 | 6 - 7 | 787 | 95 | ||
| GenBank mRNA | AB006181.1 | - | 5 - 6 | 567 | 95 | |
| AK011327.1 | - | 6 - 7 | 743 | 95 | ||
| AK156353.1 | - | 5 - 6 | 527 | 95 | ||
| BC020117.1 | - | 6 - 7 | 713 | 4009737 | 95 | |
| BC053723.1 | - | 6 - 7 | 717 | 30053020 | 95 | |
| BC085261.1 | - | 6 - 7 | 713 | 6397760 | 95 | |
| BM461330.1 | - | 4 - 5 | 418 | 95 | ||
| U62297.1 | - | 5 - 6 | 567 | 95 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nfyc
Genomic Map