Assay Details
| Gene Symbol: Ces1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081372.1 | NP_001074841.1 | 6 - 7 | 849 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ces1b| Gene Symbol: Ces1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081372.1 | NP_001074841.1 | 6 - 7 | 849 | 77 | |
Set Membership: |
|