Assay Details
| Gene Symbol: Nlrp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133946.2 | NP_598707.2 | 3 - 4 | 495 | 57 | |
| GenBank mRNA | FJ222825.1 | - | 3 - 4 | 444 | 57 | |
| JF810536.1 | - | 2 - 3 | 880 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nlrp6
Genomic Map