Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10598539 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10598529 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10598534 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10598541 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10598530 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10598544 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10598542 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10603534 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10603535 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10603536 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10598531 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10598535 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10603540 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10598545 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10603537 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10598532 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10598534 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10598531 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10598540 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10598528 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10598528 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10603538 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10598538 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10603533 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0X00011627.mm.1 |
NM_198898.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011627.mm.1 |
NM_198898.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011627.mm.1 |
TC0X00003415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000731.mm.1 |
NM_199319.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000731.mm.1 |
NM_199319.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000731.mm.1 |
TC0X00003387.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011629.mm.1 |
NM_198898.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011629.mm.1 |
NM_198898.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011629.mm.1 |
TC0X00003415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00000752.mm.1 |
NM_001081564.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000752.mm.1 |
NM_001081564.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00000752.mm.1 |
TC0X00000094.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011630.mm.1 |
NM_198898.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011630.mm.1 |
NM_198898.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011630.mm.1 |
TC0X00003415.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00011628.mm.1 |
NM_198898.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011628.mm.1 |
NM_198898.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00011628.mm.1 |
TC0X00003415.mm.1 |
MTA TC |