Assay Details
| Gene Symbol: Magea10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085506.1 | NP_001078975.1 | 2 - 3 | 158 | 107 | |
| GenBank mRNA | AK136064.1 | - | 2 - 3 | 159 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001085506.1 transcripts.
Set Membership: |
|

Magea10
Genomic Map