Assay Details
| Gene Symbol: Gm13277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098840.3 | NP_001092310.1 | 1 | 464 | 88 | |
| Gene Symbol: Gm13276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085532.3 | NP_001079001.1 | 1 | 464 | 88 | |
| Gene Symbol: Ifnz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197889.2 | NP_922871.1 | 1 | 464 | 88 | |
| GenBank mRNA | AK046550.1 | - | 1 | 465 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK046550.1, NM_001085532.3, NM_001098840.3, NM_197889.2 transcripts.
Set Membership: |
|

Gm13277
Genomic Map