Assay Details
| Gene Symbol: Krtap17-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099774.2 | NP_001093244.1 | 1 | 49 | 101 | |
| GenBank mRNA | AK020698.1 | - | 1 | 50 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001099774.2 transcripts.
Set Membership: |
|

Krtap17-1
Genomic Map