| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700020269.mm.1 |
TC0700002233.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700020269.mm.1 |
NM_001105063.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020269.mm.1 |
NM_001105063.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020678.mm.1 |
TC0700002302.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700020678.mm.1 |
NM_001105156.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020678.mm.1 |
NM_001105156.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020679.mm.1 |
TC0700002302.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700020679.mm.1 |
NM_001105156.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020679.mm.1 |
NM_001105156.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020268.mm.1 |
TC0700002233.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700020268.mm.1 |
NM_001105063.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020268.mm.1 |
NM_001105063.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020267.mm.1 |
TC0700002233.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700020267.mm.1 |
NM_001105063.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020267.mm.1 |
NM_001105063.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020677.mm.1 |
TC0700002302.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700020677.mm.1 |
NM_001105156.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700020677.mm.1 |
NM_001105156.1 |
Refseq |