Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10559876 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559900 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549908 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559889 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559877 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549924 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559886 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559873 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549942 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559872 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559929 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559885 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549927 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559874 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549944 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549911 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549905 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549909 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549941 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559884 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559897 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549939 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559875 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549907 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559888 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559899 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559898 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549910 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549925 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559930 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549943 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549906 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549940 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559896 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10549928 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10559887 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10559871 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10549938 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10559901 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10549923 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10549904 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0700020238.mm.1 |
TC0700002228.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020238.mm.1 |
NM_009490.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020238.mm.1 |
NM_009490.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020323.mm.1 |
TC0700002243.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020323.mm.1 |
NM_001105068.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020323.mm.1 |
NM_001105068.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020322.mm.1 |
TC0700002243.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020322.mm.1 |
NM_001105068.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020322.mm.1 |
NM_001105068.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020391.mm.1 |
TC0700002254.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020391.mm.1 |
NM_009493.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020391.mm.1 |
NM_009493.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020350.mm.1 |
TC0700002249.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020350.mm.1 |
NM_001105151.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020675.mm.1 |
TC0700002302.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020675.mm.1 |
NM_001105156.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020675.mm.1 |
NM_001105156.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020346.mm.1 |
TC0700002249.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020346.mm.1 |
NM_001105151.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020351.mm.1 |
TC0700002249.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020351.mm.1 |
NM_001105151.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020241.mm.1 |
TC0700002228.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020241.mm.1 |
NM_009490.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020241.mm.1 |
NM_009490.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020352.mm.1 |
TC0700002249.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020352.mm.1 |
NM_001105151.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020236.mm.1 |
TC0700002228.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020236.mm.1 |
NM_009490.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020236.mm.1 |
NM_009490.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020265.mm.1 |
TC0700002233.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020265.mm.1 |
NM_001105063.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020265.mm.1 |
NM_001105063.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020239.mm.1 |
TC0700002228.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020239.mm.1 |
NM_009490.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020239.mm.1 |
NM_009490.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020240.mm.1 |
TC0700002228.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020240.mm.1 |
NM_009490.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020240.mm.1 |
NM_009490.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020242.mm.1 |
TC0700002228.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020242.mm.1 |
NM_009490.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020242.mm.1 |
NM_009490.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020348.mm.1 |
TC0700002249.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020348.mm.1 |
NM_001105151.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020349.mm.1 |
TC0700002249.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020349.mm.1 |
NM_001105151.1 |
Refseq |
Murine Genome U74v2 Set |
97127_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
97127_f_at |
- |
- |