Assay Details
| Gene Symbol: Higd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019814.4 | NP_062788.1 | 3 | 724 | 101 | |
| GenBank mRNA | AB253941.1 | - | 2 | 236 | 101 | |
| AF141312.1 | - | 3 | 255 | 101 | ||
| AK004166.1 | - | 3 | 351 | 101 | ||
| AK008756.1 | - | 3 | 351 | 101 | ||
| AK018360.1 | - | 3 | 352 | 101 | ||
| AY028386.1 | - | 3 | 277 | 101 | ||
| BC021594.1 | - | 3 | 274 | 5148784 | 101 | |
| BC090259.1 | - | 3 | 328 | 5060413 | 101 | |
| BC132371.1 | - | 3 | 342 | 40130648 | 101 | |
| BC151191.1 | - | 3 | 342 | 8861324 | 101 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Higd1a
Genomic Map