| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100024200.mm.1 |
NM_001114679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024200.mm.1 |
NM_001114679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024200.mm.1 |
TC1100004292.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100024216.mm.1 |
NM_173434.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024216.mm.1 |
NM_173434.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024216.mm.1 |
TC1100002648.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100024217.mm.1 |
NM_173434.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024217.mm.1 |
NM_173434.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024217.mm.1 |
TC1100002648.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100024201.mm.1 |
NM_001114679.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024201.mm.1 |
NM_001114679.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024201.mm.1 |
TC1100004292.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100024215.mm.1 |
NM_173434.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024215.mm.1 |
NM_173434.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024215.mm.1 |
TC1100002648.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100024222.mm.1 |
NM_173434.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024222.mm.1 |
NM_173434.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100024222.mm.1 |
TC1100002648.mm.1 |
MTA TC |