Assay Details
Gene Symbol: Olfr1338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207152.2 | NP_997035.2 | 1 | 250 | 101 | |
GenBank mRNA | BC130245.1 | - | 1 | 254 | 40142324 | 101 |
Gene Symbol: Olfr1328 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146399.2 | NP_666511.2 | 1 | 250 | 101 | |
GenBank mRNA | BC106978.2 | - | 1 | 250 | 40042180 | 101 |
BC148290.2 | - | 1 | 250 | 40042169 | 101 |
Gene Symbol: Olfr1330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146334.2 | NP_666446.1 | 1 | 256 | 101 | |
GenBank mRNA | BC108943.1 | - | 1 | 256 | 40042936 | 101 |
BC108944.1 | - | 1 | 251 | 101 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P420457 | BC130245.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P420457 | NM_207152.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P420457 | BC130245.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P420457 | NM_207152.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150198 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P420457 | BC130245.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P420457 | NM_207152.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P420457 | BC130245.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P420457 | NM_207152.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P420457 | BC130245.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P420457 | NM_207152.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P420457 | BC130245.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P420457 | NM_207152.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242080 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242103 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242109 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242072 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242075 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242094 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242071 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242099 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242124 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242102 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242070 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242078 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242120 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242115 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242068 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242095 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242113 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242122 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242117 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242107 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23242104 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150198 | - | - |
Whole Mouse Genome G4122A | A_51_P420457 | BC130245.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P420457 | NM_207152.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P420457 | BC130245.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P420457 | NM_207152.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101122 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131476 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150198 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03392831 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03558245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23242047 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242112 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03882766 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242107 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03882767 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03317769 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242064 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242091 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242110 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242103 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03998764 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03882776 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03882774 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03998760 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242104 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242037 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03579542 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03998762 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03665015 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242060 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242095 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03593188 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242094 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242117 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242072 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242099 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242050 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242070 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242068 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242122 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242044 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03457543 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242065 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242075 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242113 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242115 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242105 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242078 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242041 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242102 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242046 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03998759 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23242059 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10515829 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515825 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10515827 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10515828 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10515824 | - | - |
Affymetrix Mouse Clariom S Array | PSR0400027608.mm.1 | TC0400003407.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400027608.mm.1 | NM_146399.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400027608.mm.1 | NM_146399.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400027593.mm.1 | TC0400003401.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400027593.mm.1 | NM_207152.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400027593.mm.1 | NM_207152.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009991.mm.1 | NM_146334.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009991.mm.1 | NM_146334.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400009991.mm.1 | TC0400001319.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5550735 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5550735 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005550735 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2769476 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2769476 | - | - |
mouse-6 v1.1 (Illumina) | scl0258259.1_104-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5550735 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0258259.1_104-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5550735 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2903964 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2769476 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2903964 | - | - |
