Assay Details
| Gene Symbol: Wrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207301.2 | NP_997184.1 | 1 - 2 | 160 | 89 | |
| GenBank mRNA | AK021091.1 | - | 1 - 2 | 159 | 89 | |
| AK172079.1 | - | 1 - 2 | 173 | 89 | ||
| BC031769.1 | - | 1 - 2 | 160 | 4235618 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wrb
Genomic Map