| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700017276.mm.1 |
NM_207541.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017276.mm.1 |
NM_207541.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017276.mm.1 |
TC1700001869.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017278.mm.1 |
NM_207541.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017278.mm.1 |
NM_207541.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017278.mm.1 |
TC1700001869.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017273.mm.1 |
NM_207541.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017273.mm.1 |
NM_207541.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017273.mm.1 |
TC1700001869.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017274.mm.1 |
NM_207541.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017274.mm.1 |
NM_207541.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017274.mm.1 |
TC1700001869.mm.1 |
MTA TC |