Assay Details
| Gene Symbol: Defa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007852.1 | NP_031878.1 | 1 - 2 | 177 | 93 | |
| Gene Symbol: Defa24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024225.2 | NP_001019396.1 | 1 - 2 | 220 | 93 | |
| GenBank mRNA | AY746429.1 | - | 1 - 2 | 177 | 93 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Whole Mouse Genome G4122A | A_52_P514200 | NM_007852.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25707360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25707383 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10570730 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570731 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570658 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570678 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570736 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570657 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570678 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570657 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570702 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570691 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570730 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570736 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570692 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570692 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570670 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570731 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570677 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570737 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570658 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570677 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570665 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570669 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570691 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570669 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10570670 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450631_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570656 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570690 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570668 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570690 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570735 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570735 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570656 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570729 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570676 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570676 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570668 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10570729 | - | - |
| Mouse Genome 430 2.0 Array | 1450631_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1450631_x_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800001927.mm.1 | NM_001024225.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001927.mm.1 | NM_001024225.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001927.mm.1 | TC0800000257.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800001930.mm.1 | NM_001024225.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001930.mm.1 | NM_001024225.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001930.mm.1 | TC0800000257.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800001932.mm.1 | NM_001024225.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001932.mm.1 | NM_001024225.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001932.mm.1 | TC0800000257.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800001929.mm.1 | NM_001024225.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001929.mm.1 | NM_001024225.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800001929.mm.1 | TC0800000257.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450631_PM_x_at | - | - |
| Mouse Expression Set 430 | 1450631_x_at | - | - |
| Murine Genome U74v2 Set | 93862_i_at | - | - |
| Murine Genome U74v2 Set | 93863_f_at | - | - |
| Murine Genome U74v2 Set | 92812_PM_f_at | - | - |
| Murine Genome U74v2 Set | 93879_PM_f_at | - | - |
| Murine Genome U74v2 Set | 93879_f_at | - | - |
| Murine Genome U74v2 Set | 93863_f_at | - | - |
| Murine Genome U74v2 Set | 92812_f_at | - | - |
| Murine Genome U74v2 Set | 93863_PM_f_at | - | - |
| Murine Genome U74v2 Set | 92812_PM_f_at | - | - |
| Murine Genome U74v2 Set | 93862_PM_i_at | - | - |
| Murine Genome U74v2 Set | 92812_f_at | - | - |
| Murine Genome U74v2 Set | 93863_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450631_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450631_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450631_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450631_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 901889 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100430075 | - | - |
| MouseWG-6 v2 Expression BeadChip | 630286 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 630286 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3710538 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100430075 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6900706 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003710538 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000630286 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006900706 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2642565 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2748943 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255021 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2748943 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255021 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254739 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2642565 | - | - |
| mouse-6 v1.1 (Illumina) | scl0013235.1_28-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_13937346-S | - | - |
| mouse-6 v1.1 (Illumina) | scl013240.1_57-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0013231.1_26-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3710538 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 630286 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6900706 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100430075 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl013240.1_57-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0013235.1_28-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0013231.1_26-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6900706 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100430075 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 630286 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3710538 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2642565 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1255021 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2642565 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254739 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255021 | - | - |

Defa6
Genomic Map