Assay Details
| Gene Symbol: Crygs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009967.2 | NP_034097.1 | 1 - 2 | 73 | 111 | |
| GenBank mRNA | AF032995.1 | - | 1 - 2 | 69 | 111 | |
| AK143060.1 | - | 1 - 2 | 70 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF032995.1, AK143060.1, NM_009967.2 transcripts.
Set Membership: |
|

Crygs
Genomic Map