Assay Details
| Gene Symbol: Olfr65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013616.4 | NP_038644.2 | 1 | 41 | 105 | |
| GenBank mRNA | BC137766.1 | - | 1 | 41 | 8860785 | 105 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC137766.1, NM_013616.4 transcripts.
Set Membership: |
|

Olfr65
Genomic Map