Assay Details
| Gene Symbol: Fxyd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022004.6 | NP_071287.1 | 3 - 4 | 398 | 113 | |
| GenBank mRNA | AA521976.1 | - | 3 - 4 | 137 | 113 | |
| AK018354.1 | - | 3 - 4 | 131 | 113 | ||
| BC042579.1 | - | 3 - 4 | 144 | 3709336 | 113 | |
| BQ891313.1 | - | 3 - 4 | 112 | 113 | ||
| W34202.1 | - | 3 - 4 | 118 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fxyd6
Genomic Map