Assay Details
| Gene Symbol: Rbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022020.2 | NP_071303.1 | 1 - 2 | 90 | 99 | |
| GenBank mRNA | AF260923.1 | - | 1 - 2 | 91 | 99 | |
| AY141971.1 | - | 1 - 2 | 100 | 99 | ||
| BC028432.1 | - | 1 - 2 | 87 | 1348253 | 99 | |
More Information
Set Membership: |
|

Rbp7
Genomic Map