Assay Details
| Gene Symbol: Asb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023046.4 | NP_075533.3 | 1 - 2 | 93 | 108 | |
| GenBank mRNA | AK006515.1 | - | 1 - 2 | 94 | 108 | |
| BC049699.1 | - | 1 - 2 | 93 | 108 | ||
| BU938650.1 | - | 1 - 2 | 96 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006515.1, NM_023046.4 transcripts.
Set Membership: |
|

Asb1
Genomic Map