Assay Details
| Gene Symbol: C1ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023143.3 | NP_075632.3 | 1 - 2 | 178 | 96 | |
| GenBank mRNA | AF148216.1 | - | 1 - 2 | 166 | 96 | |
| AI119268.1 | - | 1 - 2 | 154 | 96 | ||
| AK138817.1 | - | 1 - 2 | 77 | 96 | ||
| BC004637.1 | - | 1 - 2 | 120 | 3586055 | 96 | |
| BC093528.1 | - | 1 - 2 | 394 | 5135606 | 96 | |
| BC172078.1 | - | 1 - 2 | 103 | 9054364 | 96 | |
| BY014972.1 | - | 1 - 2 | 179 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF148216.1, AK138817.1, BC004637.1, BC093528.1, BC172078.1, NM_023143.3 transcripts.
Set Membership: |
|

C1ra
Genomic Map