Assay Details
| Gene Symbol: Dnlz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139504.1 | NP_001132976.1 | 1 - 2 | 336 | 115 | |
| GenBank mRNA | BY266852.1 | - | 1 - 2 | 303 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnlz
Genomic Map