Assay Details
| Gene Symbol: Ctnnbip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001141930.1 | NP_001135402.1 | 1 - 2 | 120 | 92 | |
| GenBank mRNA | AK003595.1 | - | 1 - 2 | 122 | 92 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK003595.1,NM_001141930.1 transcripts.
This assay is in the 5'UTR region of the AK003595.1,NM_001141930.1 transcripts.
Set Membership: |
|

Ctnnbip1
Genomic Map