Assay Details
| Gene Symbol: Gm20604 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142939.1 | NP_001136411.1 | 1 - 2 | 273 | 102 | |
| GenBank mRNA | AK011927.1 | - | 1 - 2 | 258 | 102 | |
| AK019599.1 | - | 1136 | 102 | |||
| BC014715.1 | - | 1 - 2 | 222 | 4236212 | 102 | |
| BC055374.1 | - | 1 - 2 | 226 | 30071431 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK019599.1, BC014715.1, BC055374.1 transcripts.
Set Membership: |
|

Gm20604
Genomic Map