Assay Details
| Gene Symbol: B3gnt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159407.1 | NP_001152879.1 | 2 - 3 | 433 | 72 | |
| GenBank mRNA | CN533455.1 | - | 2 - 3 | 268 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001159407.1 transcripts.
Set Membership: |
|

B3gnt5
Genomic Map