Assay Details
| Gene Symbol: Enthd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163189.1 | NP_001156661.1 | 1 - 2 | 467 | 104 | |
| GenBank mRNA | BC158112.1 | - | 1 - 2 | 467 | 9088186 | 104 |
More Information
Updated Mapping Information
Set Membership: |
|

Enthd1
Genomic Map