Assay Details
| Gene Symbol: Tcp10c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167578.1 | NP_001161050.1 | 2 - 3 | 234 | 92 | |
| GenBank mRNA | AK076614.1 | - | 2 - 3 | 180 | 92 | |
| BC061173.1 | - | 2 - 3 | 205 | 6772123 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK076614.1,BC061173.1,NM_001167578.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tcp10c
Genomic Map