Assay Details
| Gene Symbol: Eif4ebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007918.3 | NP_031944.3 | 1 - 2 | 224 | 74 | |
| GenBank mRNA | AK013033.1 | - | 150 | 74 | ||
| AK169979.1 | - | 1 - 2 | 170 | 74 | ||
| AK170031.1 | - | 1 - 2 | 174 | 74 | ||
| BC002045.1 | - | 1 - 2 | 192 | 3592595 | 74 | |
| BE651550.1 | - | 1 - 2 | 115 | 74 | ||
| BY318447.1 | - | 1 - 2 | 225 | 74 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eif4ebp1
Genomic Map