Assay Details
| Gene Symbol: G6pc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008061.3 | NP_032087.2 | 1 - 2 | 291 | 99 | |
| GenBank mRNA | AK050279.1 | - | 1 - 2 | 306 | 99 | |
| AK052656.1 | - | 1 - 2 | 306 | 99 | ||
| BC013448.1 | - | 1 - 2 | 291 | 4237115 | 99 | |
| U00445.1 | - | 1 - 2 | 306 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

G6pc
Genomic Map